1-Bromo-3,5-difluoro-4-iodobenzene 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Bromo-1,3-difluoro-2-iodobenzene
PC6449-100G
PC6449-25G
PC6449-5G
APOSPC6449-100GEA
1240
DKK
APOSPC6449-100G
APOSPC6449-25G
APOSPC6449-5G
1-Bromo-3,5-difluoro-4-iodobenzene 98%
1-Bromo-3,5-difluoro-4-iodobenzene
Formel:
C₆H₂BrF₂I Smeltepunkt: 41…42 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01861126 CAS-nummer: 160976-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...