2-Cyano-4-fluorobenzaldehyde 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Fluoro-2-formylbenzonitrile
PC6372-1G
PC6372-250MG
PC6372-5G
APOSPC6372-1GEA
1070
DKK
APOSPC6372-1G
APOSPC6372-250MG
APOSPC6372-5G
2-Cyano-4-fluorobenzaldehyde 98%
2-Cyano-4-fluorobenzaldehyde
Formel:
C₈H₄FNO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08275455 CAS-nummer: 77532-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...