Methyl3-Fluor-4-methoxybenzoat
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 3-fluoro-4-methoxybenzoate, 3-Fluoro-4-methoxybenzoic acid methyl ester
PC6363-100G
PC6363-25G
PC6363-5G
APOSPC6363-100GEA
2360
DKK
APOSPC6363-100G
APOSPC6363-25G
APOSPC6363-5G
Methyl3-Fluor-4-methoxybenzoat
Methyl3-Fluor-4-methoxybenzoat
Formel:
C₉H₉FO₃ Molekylvægt: 184.17 g/mol Smeltepunkt: 69…73 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00215839 CAS-nummer: 369-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...