Perfluortetradecan
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC6216-1G
PC6216-5G
APOSPC6216-1GEA
833
DKK
APOSPC6216-1G
APOSPC6216-5G
Perfluortetradecan
Perfluortetradecan
Formel:
C₁₄F₃₀ Smeltepunkt: 103…105 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013571 CAS-nummer: 307-62-0 EINECS: 206-206-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...