4-Bromo-2-(trifluoromethyl)benzoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC6177-1G PC6177-5G PC6177-25G
APOSPC6177-1GEA 164 DKK
APOSPC6177-1G APOSPC6177-5G APOSPC6177-25G
4-Bromo-2-(trifluoromethyl)benzoic acid
4-Bromo-2-(trifluoromethyl)benzoic acid
Formel: C₈H₄BrF₃O₂
Smeltepunkt: 116…121 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07787468
CAS-nummer: 320-31-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR