1-Bromperfluorhexan 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC6085W-25G
PC6085W-5G
PC6085W-100G
APOSPC6085W-25GEA
1090
DKK
APOSPC6085W-25G
APOSPC6085W-5G
APOSPC6085W-100G
1-Bromperfluorhexan 98%
1-Bromperfluorhexan
Formel:
BrF₂C(CF₂)₄CF₃ Kogepunkt: 97…101 °C Massefylde: 1,871 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00042349 CAS-nummer: 335-56-8 EINECS: 206-391-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...