4-Fluoro-2-methylbenzenesulphonamide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC5407-1G
PC5407-5G
PC5407-25G
APOSPC5407-1GEA
218
DKK
APOSPC5407-1G
APOSPC5407-5G
APOSPC5407-25G
4-Fluoro-2-methylbenzenesulphonamide
4-Fluoro-2-methylbenzenesulphonamide
Formel:
C₇H₈FNO₂S Molekylvægt: 189.2073 g/mol Smeltepunkt: 179…184 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02709831 CAS-nummer: 489-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...