2-Fluoro-4-nitro-1-(2,2,2-trifluoroethyl)benzene 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC520740-10G PC520740-1G PC520740-5G
APOSPC520740-10GEA 10460 DKK
APOSPC520740-10G APOSPC520740-1G APOSPC520740-5G
2-Fluoro-4-nitro-1-(2,2,2-trifluoroethyl)benzene 95%
2-Fluoro-4-nitro-1-(2,2,2-trifluoroethyl)benzene
Formel: C₈H₅F₄NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18384835
CAS-nummer: 1262412-94-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR