3-Chlor-4-Fluorbenzensulfonamid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC520504-25G PC520504-5G
APOSPC520504-25GEA 4370 DKK
APOSPC520504-25G APOSPC520504-5G
3-Chlor-4-Fluorbenzensulfonamid 97%
3-Chlor-4-Fluorbenzensulfonamid
Formel: C₆H₅ClFNO₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00130211
CAS-nummer: 146533-46-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR