3-Fluor-5-methoxybenzonitril 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC5077-1G
PC5077-25G
PC5077-5G
APOSPC5077-1GEA
164
DKK
APOSPC5077-1G
APOSPC5077-25G
APOSPC5077-5G
3-Fluor-5-methoxybenzonitril 98%
3-Fluor-5-methoxybenzonitril
Formel:
C₈H₆FNO Smeltepunkt: 98…100 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06797883 CAS-nummer: 439280-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...