Methyl-2-bromo-3-[4-(trifluoromethoxy)phenyl]propionate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC5069-1G PC5069-250MG
APOSPC5069-1GEA 458 DKK
APOSPC5069-1G APOSPC5069-250MG
Methyl-2-bromo-3-[4-(trifluoromethoxy)phenyl]propionate 98%
Methyl-2-bromo-3-[4-(trifluoromethoxy)phenyl]propionate
Formel: C₁₁H₁₀BrF₃O₃
Molekylvægt: 327.1 g/mol
Kogepunkt: 100…104 °C/0.8mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00219777
CAS-nummer: 175203-81-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR