(4-Bromo-2-(trifluoromethoxy)phenyl)methanol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Bromo-2-(trifluoromethoxy)benzyl alcohol

PC502690-1G PC502690-5G
APOSPC502690-1GEA 644 DKK
APOSPC502690-1G APOSPC502690-5G
(4-Bromo-2-(trifluoromethoxy)phenyl)methanol
(4-Bromo-2-(trifluoromethoxy)phenyl)methanol
Formel: C₈H₆BrF₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12546981
CAS-nummer: 220996-81-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR