2,4-Dichloro-6-fluoroquinazoline 97+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC502106-1G
PC502106-5G
APOSPC502106-1GEA
775
DKK
APOSPC502106-1G
APOSPC502106-5G
2,4-Dichloro-6-fluoroquinazoline 97+%
2,4-Dichloro-6-fluoroquinazoline
Formel:
C₈H₃Cl₂FN₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09954884 CAS-nummer: 134517-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...