Kaliumcyclopropyltrifluorborat 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC50056-25G PC50056-100G PC50056-5G
APOSPC50056-25GEA 545 DKK
APOSPC50056-25G APOSPC50056-100G APOSPC50056-5G
Kaliumcyclopropyltrifluorborat 96%
Kaliumcyclopropyltrifluorborat
Formel: C₃H₅BF₃K
Smeltepunkt: 348…350 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09265031
CAS-nummer: 1065010-87-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR