4-Bromo-2-iodo-1-(trifluoromethyl)benzene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC500392-1G PC500392-5G
APOSPC500392-1GEA 162 DKK
APOSPC500392-1G APOSPC500392-5G
4-Bromo-2-iodo-1-(trifluoromethyl)benzene
4-Bromo-2-iodo-1-(trifluoromethyl)benzene
Formel: C₇H₃BrF₃I
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD21603817
CAS-nummer: 1256945-00-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR