2,6-Difluoro-3-(trifluoromethyl)benzoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC500239-1G PC500239-5G PC500239-250MG
APOSPC500239-1GEA 741 DKK
APOSPC500239-1G APOSPC500239-5G APOSPC500239-250MG
2,6-Difluoro-3-(trifluoromethyl)benzoic acid
2,6-Difluoro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
Molekylvægt: 226.1 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03412249
CAS-nummer: 1048921-49-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR