1-Bromo-2,3-difluoro-4-nitrobenzene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC500190-1G PC500190-5G
APOSPC500190-1GEA 280 DKK
APOSPC500190-1G APOSPC500190-5G
1-Bromo-2,3-difluoro-4-nitrobenzene
1-Bromo-2,3-difluoro-4-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09839215
CAS-nummer: 1003708-24-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR