(2-Chlor-3-(trifluormethyl)phenyl)eddikesyre 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC49646-1G
PC49646-5G
APOSPC49646-1GEA
426
DKK
APOSPC49646-1G
APOSPC49646-5G
(2-Chlor-3-(trifluormethyl)phenyl)eddikesyre 97%
(2-Chlor-3-(trifluormethyl)phenyl)eddikesyre
Formel:
C₉H₆ClF₃O₂ Smeltepunkt: 131…133 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04115992 CAS-nummer: 22902-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...