2-Bromo-5-fluoropyrimidine 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-bromo-5-fluoropyrimidine
PC49168-1G
PC49168-250MG
APOSPC49168-1GEA
458
DKK
APOSPC49168-1G
APOSPC49168-250MG
2-Bromo-5-fluoropyrimidine 97%
2-Bromo-5-fluoropyrimidine
Formel:
C₄H₂BrFN₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09835164 CAS-nummer: 947533-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...