1-Bromo-5-fluoropentane

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC49114-1G PC49114-5G
APOSPC49114-1GEA 985 DKK
APOSPC49114-1G APOSPC49114-5G
1-Bromo-5-fluoropentane
1-Bromo-5-fluoropentane
Formel: C₅H₁₀BrF
Molekylvægt: 169.037 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01709395
CAS-nummer: 407-97-6
UN nummer: 1993
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR