Methyl-5-fluoro-2-iodobenzoate 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC48322-1G
PC48322-5G
PC48322-10G
APOSPC48322-1GEA
164
DKK
APOSPC48322-1G
APOSPC48322-5G
APOSPC48322-10G
Methyl-5-fluoro-2-iodobenzoate 97%
Methyl-5-fluoro-2-iodobenzoate
Formel:
C₈H₆FIO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD12965043 CAS-nummer: 1202897-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...