Hexafluoro-2-(p-tolyl)isopropanol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1,1,3,3,3-Hexafluoro-2-(4-methylphenyl)propan-2-ol

PC4805-100G PC4805-25G
APOSPC4805-100GEA 5210 DKK
APOSPC4805-100G APOSPC4805-25G
Hexafluoro-2-(p-tolyl)isopropanol 97%
Hexafluoro-2-(p-tolyl)isopropanol
Formel: C₁₀H₈F₆O
Molekylvægt: 258.17 g/mol
Kogepunkt: 182 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015357
CAS-nummer: 2010-61-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR