2-Chloromethyl-6-fluoro-1H-benzoimidazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(Chloromethyl)-6-fluoro-1H-benzo[d]imidazole

PC446031-1G PC446031-5G
APOSPC446031-1GEA 1260 DKK
APOSPC446031-1G APOSPC446031-5G
2-Chloromethyl-6-fluoro-1H-benzoimidazole
2-Chloromethyl-6-fluoro-1H-benzoimidazole
Formel: C₈H₆ClFN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08234909
CAS-nummer: 156144-42-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR