3-Bromo-2-fluoro-5-nitropyridine 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC445025-1G PC445025-25G PC445025-5G
APOSPC445025-1GEA 382 DKK
APOSPC445025-1G APOSPC445025-25G APOSPC445025-5G
3-Bromo-2-fluoro-5-nitropyridine 97%
3-Bromo-2-fluoro-5-nitropyridine
Formel: C₅H₂BrFN₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08703668
CAS-nummer: 1868-58-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR