4'-Fluorpropiophenon 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC4190-100G
PC4190-25G
APOSPC4190-100GEA
545
DKK
APOSPC4190-100G
APOSPC4190-25G
4'-Fluorpropiophenon 98%
Formel:
C₉H₉FO Kogepunkt: 108 °C/18mmHg Massefylde: 1,096 g/cm³ (25 °C) Flammepunkt: 76 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000356 CAS-nummer: 456-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...