1-(4-fluorphenyl)imidazol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-(4-Fluorophenyl)-1H-imidazole
PC4158F-10G
PC4158F-2G
APOSPC4158F-10GEA
4920
DKK
APOSPC4158F-10G
APOSPC4158F-2G
1-(4-fluorphenyl)imidazol
1-(4-fluorphenyl)imidazol
Formel:
C₉H₇FN₂ Kogepunkt: 80 °C/0,3mmHg Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00060490 CAS-nummer: 21441-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...