(4-fluorphenyl)eddikesyre 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Fluorophenyl)acetic acid

PC4140-100G PC4140-25G PC4140-500G
APOSPC4140-100GEA 230 DKK
APOSPC4140-100G APOSPC4140-25G APOSPC4140-500G
(4-fluorphenyl)eddikesyre 97%
(4-fluorphenyl)eddikesyre
Formel: (C₆H₄F)CH₂COOH
Kogepunkt: 164 °C/2mmHg
Smeltepunkt: 81…85 °C
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004343
CAS-nummer: 405-50-5
EINECS: 206-972-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR