5-Fluoronaphthalen-1-ol 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC409513-1G PC409513-5G PC409513-100MG
APOSPC409513-1GEA 6830 DKK
APOSPC409513-1G APOSPC409513-5G APOSPC409513-100MG
5-Fluoronaphthalen-1-ol 95+%
5-Fluoronaphthalen-1-ol
Formel: C₁₀H₇FO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12401609
CAS-nummer: 61761-32-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR