Dimethyl difluoromalonate 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC408111-1G
PC408111-25G
PC408111-5G
APOSPC408111-1GEA
241
DKK
APOSPC408111-1G
APOSPC408111-25G
APOSPC408111-5G
Dimethyl difluoromalonate 98%
Dimethyl difluoromalonate
Formel:
C₅H₆F₂O₄ Kogepunkt: 167…169 °C Massefylde: 1,3059 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00516931 CAS-nummer: 379-95-3 UN nummer: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...