(Nonafluorbutyl)undecafluorcyclohexan

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC408080-25G PC408080-5G
APOSPC408080-25GEA 384 DKK
APOSPC408080-25G APOSPC408080-5G
(Nonafluorbutyl)undecafluorcyclohexan
(Nonafluorobutyl)undecafluorocyclohexane
Formel: C₁₀F₂₀
Kogepunkt: 145…147 °C
Massefylde: 1,899 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 374-60-7
EINECS: 206-777-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR