2-Amino-4-(trifluormethyl)pyridin
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC4027-25G
PC4027-5G
APOSPC4027-25GEA
676
DKK
APOSPC4027-25G
APOSPC4027-5G
2-Amino-4-(trifluormethyl)pyridin
2-Amino-4-(trifluoromethyl)pyridine
Formel:
C₆H₅F₃N₂ Smeltepunkt: 70…74 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00077455 CAS-nummer: 106447-97-6 UN nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...