3-(3,5-Difluorphenyl)propionsyre 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC400710-5G
PC400710-10G
APOSPC400710-5GEA
469
DKK
APOSPC400710-5G
APOSPC400710-10G
3-(3,5-Difluorphenyl)propionsyre 97%
3-(3,5-Difluorphenyl)propionsyre
Formel:
C₉H₈F₂O₂ Smeltepunkt: 60 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04116054 CAS-nummer: 84315-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...