Bis(3-fluorphenyl)disulfid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC3979-25G
PC3979-5G
APOSPC3979-25GEA
1530
DKK
APOSPC3979-25G
APOSPC3979-5G
Bis(3-fluorphenyl)disulfid 97%
Bis(3-fluorphenyl)disulfid
Formel:
C₁₂H₈F₂S₂ Kogepunkt: 308 °C Massefylde: 1,27 g/cm³ (25 °C) Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01318104 CAS-nummer: 63930-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...