2-Fluor-4-nitroanisol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC3882-100G PC3882-25G PC3882-5G
APOSPC3882-100GEA 436 DKK
APOSPC3882-100G APOSPC3882-25G APOSPC3882-5G
2-Fluor-4-nitroanisol 98%
2-Fluor-4-nitroanisol
Formel: C₇H₆FNO₃
Molekylvægt: 171.13 g/mol
Smeltepunkt: 103…105 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00061095
CAS-nummer: 455-93-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR