6-Fluorquinaldin 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC3827-1G PC3827-5G
APOSPC3827-1GEA 164 DKK
APOSPC3827-1G APOSPC3827-5G
6-Fluorquinaldin 97%
6-Fluorquinaldin
Formel: C₁₀H₈FN
Smeltepunkt: 50…54 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041233
CAS-nummer: 1128-61-6
EINECS: 214-439-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR