3-Fluor-6-methylbenzoesyre 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC3823BM-100G PC3823BM-25G PC3823BM-5G
APOSPC3823BM-100GEA 458 DKK
APOSPC3823BM-100G APOSPC3823BM-25G APOSPC3823BM-5G
3-Fluor-6-methylbenzoesyre 98%
3-Fluor-6-methylbenzoesyre
Formel: C₈H₇FO₂
Smeltepunkt: 128…132 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042294
CAS-nummer: 33184-16-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR