3-Fluoro-4-methylbenzenecarboximidamide hydrochloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Fluoro-4-methylbenzimidamide hydrochloride, 3-Fluoro-4-methylbenzamidine hydrochloride

PC3822J-1G PC3822J-5G
APOSPC3822J-1GEA 872 DKK
APOSPC3822J-1G APOSPC3822J-5G
3-Fluoro-4-methylbenzenecarboximidamide hydrochloride 97%
3-Fluoro-4-methylbenzenecarboximidamide hydrochloride
Formel: C₈H₁₀ClFN₂
Molekylvægt: 188.63 g/mol
Smeltepunkt: 165…167 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00275569
CAS-nummer: 175277-88-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR