4-Fluoro-4'-methoxybenzophenone 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(4-Fluorophenyl)(4-methoxyphenyl)methanone
PC3817FM-25G
PC3817FM-100G
PC3817FM-500G
APOSPC3817FM-25GEA
208
DKK
APOSPC3817FM-25G
APOSPC3817FM-100G
APOSPC3817FM-500G
4-Fluoro-4'-methoxybenzophenone 99%
4-Fluoro-4'-methoxybenzophenone
Formel:
C₁₄H₁₁FO₂ Smeltepunkt: 90…92 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00055469 CAS-nummer: 345-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...