Poly(chlorotrifluoroethylen) 800

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC3814B-1LB PC3814B-5LB
APOSPC3814B-1LBEA 7650 DKK
APOSPC3814B-1LB APOSPC3814B-5LB
Poly(chlorotrifluoroethylen) 800
Poly(chlorotrifluoroethylen) 800
Formel: (C₂ClF₃)n
Molekylvægt: 800 g/mol
Massefylde: 1.9 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 9002-83-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR