2-(Trifluoromethyl)dioxolane 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC3398-5G
PC3398-1G
APOSPC3398-5GEA
2680
DKK
APOSPC3398-5G
APOSPC3398-1G
2-(Trifluoromethyl)dioxolane 97%
2-(Trifluoromethyl)dioxolane
Formel:
C₄H₅F₃O₂ Kogepunkt: 90 °C Massefylde: 1,46 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04038366 CAS-nummer: 2344-09-4 UN nummer: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...