2-Fluoranilin 99+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC3390-1KG
PC3390-250G
PC3390-500G
APOSPC3390-1KGEA
622
DKK
APOSPC3390-1KG
APOSPC3390-250G
APOSPC3390-500G
2-Fluoranilin 99+%
2-Fluoranilin
Intermediate for the synthesis of Flurbiprofen, an important NSAID.
Formel:
C₆H₆FN Molekylvægt: 111.12 g/mol Kogepunkt: 182…183 °C Smeltepunkt: –29 °C Massefylde: 1.151 g/cm³ (25 °C) Flammepunkt: 60 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007642 CAS-nummer: 348-54-9 EINECS: 206-478-9 UN nummer: 2941 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...