3,4,5-Trifluorophenylacetyl chloride
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC32866-1G
PC32866-250MG
PC32866-500MG
APOSPC32866-1GEA
3440
DKK
APOSPC32866-1G
APOSPC32866-250MG
APOSPC32866-500MG
3,4,5-Trifluorophenylacetyl chloride
3,4,5-Trifluorophenylacetyl chloride
Formel:
C₈H₄ClF₃O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD12025027 CAS-nummer: 849623-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...