2-Fluoro-3-formylphenylboronic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Fluoro-3-formylbenzeneboronic acid, (2-Fluoro-3-formylphenyl)boronic acid

PC32756-100G PC32756-25G PC32756-5G
APOSPC32756-100GEA 4130 DKK
APOSPC32756-100G APOSPC32756-25G APOSPC32756-5G
2-Fluoro-3-formylphenylboronic acid
2-Fluoro-3-formylphenylboronic acid
Formel: C₇H₆BFO₃
Smeltepunkt: 100…105 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07369725
CAS-nummer: 849061-98-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR