2-(Bromodifluoromethyl)benzoxazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC32712-1G
PC32712-5G
APOSPC32712-1GEA
654
DKK
APOSPC32712-1G
APOSPC32712-5G
2-(Bromodifluoromethyl)benzoxazole
2-(Bromodifluoromethyl)benzoxazole
Formel:
C₈H₄BrF₂NO Kogepunkt: 62 °C/1.5mmHg Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11035866 CAS-nummer: 186828-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...