Ethyl-5-(4-chlorophenyl)-2-(trifluoromethyl)-3-furoate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC3183-1G
PC3183-5G
APOSPC3183-1GEA
985
DKK
APOSPC3183-1G
APOSPC3183-5G
Ethyl-5-(4-chlorophenyl)-2-(trifluoromethyl)-3-furoate
Ethyl-5-(4-chlorophenyl)-2-(trifluoromethyl)-3-furoate
Formel:
C₁₄H₁₀ClF₃O₃ Molekylvægt: 318.68 g/mol Smeltepunkt: 91…93 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00275556 CAS-nummer: 175276-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...