5-Fluoro-2-methyl-1H-indene-3-acetic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (5-Fluoro-2-methyl-1H-inden-3-yl)acetic acid

PC3174-1G PC3174-500MG
APOSPC3174-1GEA 472 DKK
APOSPC3174-1G APOSPC3174-500MG
5-Fluoro-2-methyl-1H-indene-3-acetic acid
5-Fluoro-2-methyl-1H-indene-3-acetic acid
Formel: C₁₂H₁₁FO₂
Smeltepunkt: 161…163 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01076194
CAS-nummer: 32004-66-3
EINECS: 250-891-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR