2-Chloro-4-(trifluoromethyl)benzyl bromide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC303117-1G PC303117-5G PC303117-10G
APOSPC303117-1GEA 1530 DKK
APOSPC303117-1G APOSPC303117-5G APOSPC303117-10G
2-Chloro-4-(trifluoromethyl)benzyl bromide 98%
2-Chloro-4-(trifluoromethyl)benzyl bromide
Formel: C₈H₅BrClF₃
Molekylvægt: 273.48 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14155638
CAS-nummer: 279252-26-5
UN nummer: 3265
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR