3,5-Difluoro-2-methoxyaniline 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC303052-5G PC303052-1G
APOSPC303052-5GEA 1160 DKK
APOSPC303052-5G APOSPC303052-1G
3,5-Difluoro-2-methoxyaniline 98%
3,5-Difluoro-2-methoxyaniline
Formel: C₇H₇F₂NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09832386
CAS-nummer: 41860-67-7
UN nummer: 2810
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR