2-Methyl-5-(pentafluorothio)benzyl bromide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Methyl-5-(pentafluorosulphur)benzyl bromide

PC303046-1G PC303046-5G
APOSPC303046-1GEA 2620 DKK
APOSPC303046-1G APOSPC303046-5G
2-Methyl-5-(pentafluorothio)benzyl bromide 98%
2-Methyl-5-(pentafluorothio)benzyl bromide
Formel: C₈H₈BrF₅S
Molekylvægt: 311.11 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16652479
CAS-nummer: 1240257-10-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR