(2,3-Difluor-6-methoxyphenyl)eddikesyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC302374-1G
PC302374-5G
APOSPC302374-1GEA
1220
DKK
APOSPC302374-1G
APOSPC302374-5G
(2,3-Difluor-6-methoxyphenyl)eddikesyre
(2,3-Difluor-6-methoxyphenyl)eddikesyre
Formel:
(C₆H₂F₂(OCH₃))CH₂COOH Smeltepunkt: 125…127 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04116025 CAS-nummer: 886501-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...